1-Chlorooctane ≥99%

Supplier: Ambeed
Danger

A1453658-25G A1453658-100G
76661-594EA 14 USD
76661-594 77874-926
1-Chlorooctane ≥99%
1-Chlorooctane

Precaución: for Research Use Only

Formula: C₈H₁₇Cl
MDL Number: MFCD00001022
CAS Number: 111-85-3
UN: 3082
ADR: 9,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR