Succinic acid 99.5%

Supplier: Ambeed
Warning

Synonyms: 1,4-Butanedioic acid

A1453553-1000G A1453553-100G A1453553-25G A1453553-500G
76661-578EA 21 USD
76661-578 76661-580 76661-582 76661-584
Succinic acid 99.5%
Succinic acid

Precaución: for Research Use Only

Formula: HOOCCH₂CH₂COOH
MW: 118.09 g/mol
Boiling Pt: 235
Melting Pt: 182
Flash Pt: 206
MDL Number: MFCD00002789
CAS Number: 110-15-6
Merck Index: 13,08953

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR