2-Methoxy-4-nitrobenzonitrile 97% (by GC)

Supplier: Ambeed
Warning

A1453230-1G A1453230-25G A1453230-5G
76661-562EA 20.16 USD
76661-562 76661-564 76661-566
2-Methoxy-4-nitrobenzonitrile 97% (by GC)
2-Methoxy-4-nitrobenzonitrile

Precaución: for Research Use Only

Formula: C₈H₆N₂O₃
MW: 178.15 g/mol
MDL Number: MFCD02093781
CAS Number: 101084-96-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR