(±)-2,2-Dimethyl-1,3-dioxolan-4-ylmethanol 99% contains isomer
Supplier: Ambeed
|
Warning
|
Synonyms:
(2,2-Dimethyl-1,3-dioxolan-4-yl)methanol, 2,2-Dimethyl-1,3-dioxolan-4-ylmethanol, DL-1,2-Isopropylidenglycerol, 1,2-O-Isopropylideneglycerol, Solketal, (±)-1,2-O-Isopropylidenglycerol, DL-1,2-Isopropylideneglycerol
A1453188-100G
A1453188-25G
A1453188-500G
A1453188-1000G
76661-556EA
9.55
USD
76661-556
76661-558
76661-560
77566-934
(±)-2,2-Dimethyl-1,3-dioxolan-4-ylmethanol 99% contains isomer
(±)-2,2-Dimethyl-1,3-dioxolan-4-ylmethanol
Precaución: For research use only.
Formula:
C₆H₁₂O₃ MW: 132.16 g/mol |
MDL Number:
MFCD00063238 CAS Number: 100-79-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...