Allylamine hydrochloride 60% in water

Supplier: Ambeed
Danger

Synonyms: Allylamine Hydrochloride

A1453073-25G A1453073-100G
76661-548EA 8 USD
76661-548 77874-921
Allylamine hydrochloride 60% in water
Allylamine hydrochloride

Precaución: for Research Use Only

Formula: C₃H₈ClN
MDL Number: MFCD00054329
CAS Number: 10017-11-5
UN: 2811
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR