2-Bromophenyl trifluoromethanesulfonate 97%

Supplier: Ambeed
Warning

Synonyms: 2-Bromophenyl triflate, Trifluoromethanesulphonic acid 2-Bromophenyl ester

A1453031-1G A1453031-250MG A1453031-25G A1453031-5G
76661-540EA 18 USD
76661-540 76661-542 76661-544 76661-546
2-Bromophenyl trifluoromethanesulfonate 97%
2-Bromophenyl trifluoromethanesulfonate

Precaución: for Research Use Only

Formula: C₇H₄BrF₃O₃S
MW: 305.07 g/mol
MDL Number: MFCD18379777
CAS Number: 129112-25-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR