2-(4-(Dimethylamino)phenyl)benzo[d]isothiazol-3(2H)-one 97%

Supplier: Ambeed
Warning

A1453017-100MG A1453017-1G A1453017-250MG
76661-530EA 56 USD
76661-530 76661-532 76661-534
2-(4-(Dimethylamino)phenyl)benzo[d]isothiazol-3(2H)-one 97%
2-(4-(Dimethylamino)phenyl)benzo[d]isothiazol-3(2H)-one

Precaución: for Research Use Only

Formula: C₁₅H₁₄N₂OS
CAS Number: 106595-93-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR