N-(2-Chloroethyl)-3-cyanobenzamide 97%

Supplier: Ambeed
Warning

A1452955-1G A1452955-5G
76661-526EA 29.71 USD
76661-526 76661-528
N-(2-Chloroethyl)-3-cyanobenzamide 97%
N-(2-Chloroethyl)-3-cyanobenzamide

Precaución: for Research Use Only

Formula: C₁₀H₉ClN₂O
MDL Number: MFCD14613726
CAS Number: 1247759-33-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR