1,3,5-Tris(1-phenyl-1H-benzimidazol-2-yl)benzene 99.9%, purified by sublimation

Supplier: Ambeed
Warning

Synonyms: 2,2',2''-(1,3,5-Benzenetriyl)tris(1-phenyl-1H-benzimidazole), TPBi, 1,3,5-Tris(1-phenyl-1H-benzo[d]imidazol-2-yl)benzene

A1452912-100MG A1452912-250MG A1452912-5G
76661-520EA 30.77 USD
76661-520 76661-522 76661-524
1,3,5-Tris(1-phenyl-1H-benzimidazol-2-yl)benzene 99.9%, purified by sublimation
1,3,5-Tris(1-phenyl-1H-benzimidazol-2-yl)benzene

Precaución: for Research Use Only

Formula: C₄₅H₃₀N₆
MW: 654.78 g/mol
MDL Number: MFCD08276433
CAS Number: 192198-85-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR