N,N-Diethyl-4-((trimethylsilyl)ethynyl)aniline 97%

Supplier: Ambeed
Warning

A1452893-100MG A1452893-1G A1452893-250MG
76661-514EA 142.17 USD
76661-514 76661-516 76661-518
N,N-Diethyl-4-((trimethylsilyl)ethynyl)aniline 97%
N,N-Diethyl-4-((trimethylsilyl)ethynyl)aniline

Precaución: for Research Use Only

Formula: C₁₅H₂₃NSi
MW: 245.4353 g/mol
CAS Number: 562813-15-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR