3-(7-Bromo-4-fluoro-1-oxoisoindolin-2-yl)piperidine-2,6-dione 98%

Supplier: Ambeed
Warning

A1452500-100MG A1452500-1G A1452500-250MG A1452500-50MG
76661-502EA 26 USD
76661-502 76661-504 76661-506 76661-508
3-(7-Bromo-4-fluoro-1-oxoisoindolin-2-yl)piperidine-2,6-dione 98%
3-(7-Bromo-4-fluoro-1-oxoisoindolin-2-yl)piperidine-2,6-dione

Precaución: for Research Use Only

Formula: C₁₃H₁₀BrFN₂O₃
CAS Number: 2438243-28-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR