3-(4-Bromo-7-fluoro-1-oxoisoindolin-2-yl)piperidine-2,6-dione 98%

Supplier: Ambeed
Warning

A1452499-100MG A1452499-1G A1452499-250MG A1452499-50MG
76661-494EA 27 USD
76661-494 76661-496 76661-498 76661-500
3-(4-Bromo-7-fluoro-1-oxoisoindolin-2-yl)piperidine-2,6-dione 98%
3-(4-Bromo-7-fluoro-1-oxoisoindolin-2-yl)piperidine-2,6-dione

Precaución: for Research Use Only

Formula: C₁₃H₁₀BrFN₂O₃
CAS Number: 2438239-34-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR