2-(4-(Trifluoromethyl)phenyl)pyrimidin-4-amine 97%

Supplier: Ambeed
Warning

A1452478-100MG A1452478-1G A1452478-250MG
76661-488EA 117.76 USD
76661-488 76661-490 76661-492
2-(4-(Trifluoromethyl)phenyl)pyrimidin-4-amine 97%
2-(4-(Trifluoromethyl)phenyl)pyrimidin-4-amine

Precaución: for Research Use Only

Formula: C₁₁H₈F₃N₃
MW: 239.2000 g/mol
MDL Number: MFCD11045645
CAS Number: 1215072-94-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR