1-Methyl-4-(piperidin-4-ylmethyl)piperazine hydrochloride 97%

Supplier: Ambeed
Warning

A1452472-100MG A1452472-250MG A1452472-5G
76661-482EA 29.71 USD
76661-482 76661-484 76661-486
1-Methyl-4-(piperidin-4-ylmethyl)piperazine hydrochloride 97%
1-Methyl-4-(piperidin-4-ylmethyl)piperazine hydrochloride

Precaución: for Research Use Only

Formula: C₁₁H₂₄ClN₃
MW: 233.7800 g/mol
CAS Number: 1831151-60-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR