(5-(((tert-Butyldimethylsilyl)oxy)methyl)-1-methyl-1H-pyrazol-3-yl)methanol 97%

Supplier: Ambeed
Warning

A1452451-100MG A1452451-1G A1452451-250MG
76661-476EA 63.66 USD
76661-476 76661-478 76661-480
(5-(((tert-Butyldimethylsilyl)oxy)methyl)-1-methyl-1H-pyrazol-3-yl)methanol 97%
(5-(((tert-Butyldimethylsilyl)oxy)methyl)-1-methyl-1H-pyrazol-3-yl)methanol

Precaución: for Research Use Only

Formula: C₁₂H₂₄N₂O₂Si
MW: 256.4167 g/mol
CAS Number: 2450998-49-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR