2-(2-Bromo-3-chloro-4-fluorophenyl)acetic acid 97%

Supplier: Ambeed
Warning

A1452411-100MG A1452411-250MG
76661-470EA 108.22 USD
76661-470 76661-472
2-(2-Bromo-3-chloro-4-fluorophenyl)acetic acid 97%
2-(2-Bromo-3-chloro-4-fluorophenyl)acetic acid

Precaución: for Research Use Only

Formula: C₈H₅BrClFO₂
MW: 267.4795 g/mol
CAS Number: 2384318-64-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR