(SP-5-13)-Chloro[[2,2'-[1,2-ethanediylbis[(nitrilo-κN)methylidyne]]bis[4,6-bis(1,1-dimethylethyl)phenolato-κO]](2-)]manganese 97%

Supplier: Ambeed
Warning

A1452135-10G A1452135-1G A1452135-5G
76661-428EA 506.05 USD
76661-428 76661-430 76661-432
(SP-5-13)-Chloro[[2,2'-[1,2-ethanediylbis[(nitrilo-κN)methylidyne]]bis[4,6-bis(1,1-dimethylethyl)phenolato-κO]](2-)]manganese 97%
(SP-5-13)-Chloro[[2,2'-[1,2-ethanediylbis[(nitrilo-κN)methylidyne]]bis[4,6-bis(1,1-dimethylethyl)phenolato-κO]](2-)]manganese

Precaución: for Research Use Only

Formula: C₃₂H₄₆ClMnN₂O₂
MW: 581.1109 g/mol
CAS Number: 172172-24-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR