6,6'-((Ethane-1,2-diylbis(azanylylidene))bis(methanylylidene))bis(2,4-di-tert-butylphenol) 97%

Supplier: Ambeed
Warning

A1452131-100G A1452131-10G A1452131-1G A1452131-25G A1452131-5G
76661-420EA 1178.66 USD
76661-420 76661-422 76661-424 76661-426 77597-224
6,6'-((Ethane-1,2-diylbis(azanylylidene))bis(methanylylidene))bis(2,4-di-tert-butylphenol) 97%
6,6'-((Ethane-1,2-diylbis(azanylylidene))bis(methanylylidene))bis(2,4-di-tert-butylphenol)

Precaución: for Research Use Only

Formula: C₃₂H₄₈N₂O₂
MW: 492.7357 g/mol
MDL Number: MFCD00312332
CAS Number: 103595-81-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR