2,2-Bis((R)-4-(tert-butyl)-4,5-dihydrooxazol-2-yl)acetonitrile 97%

Supplier: Ambeed
Warning

A1451998-100MG A1451998-1G A1451998-250MG
76661-400EA 150.65 USD
76661-400 76661-402 76661-404
2,2-Bis((R)-4-(tert-butyl)-4,5-dihydrooxazol-2-yl)acetonitrile 97%
2,2-Bis((R)-4-(tert-butyl)-4,5-dihydrooxazol-2-yl)acetonitrile

Precaución: for Research Use Only

Formula: C₁₆H₂₅N₃O₂
MW: 291.3800 g/mol
CAS Number: 1442644-13-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR