2,2-Bis((S)-4-(tert-butyl)-4,5-dihydrooxazol-2-yl)acetonitrile 97%

Supplier: Ambeed
Warning

A1451997-100MG A1451997-1G A1451997-250MG
76661-394EA 103.97 USD
76661-394 76661-396 76661-398
2,2-Bis((S)-4-(tert-butyl)-4,5-dihydrooxazol-2-yl)acetonitrile 97%
2,2-Bis((S)-4-(tert-butyl)-4,5-dihydrooxazol-2-yl)acetonitrile

Precaución: for Research Use Only

CAS Number: 1251471-84-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR