2-(2,6-Dioxopiperidin-3-yl)-4-(prop-2-yn-1-ylamino)isoindoline-1,3-dione 97%

Supplier: Ambeed
Warning

A1448901-100MG A1448901-1G A1448901-250MG A1448901-5G
76661-366EA 51 USD
76661-366 76661-368 76661-370 77622-152
2-(2,6-Dioxopiperidin-3-yl)-4-(prop-2-yn-1-ylamino)isoindoline-1,3-dione 97%
2-(2,6-Dioxopiperidin-3-yl)-4-(prop-2-yn-1-ylamino)isoindoline-1,3-dione

Precaución: for Research Use Only

Formula: C₁₆H₁₃N₃O₄
MDL Number: MFCD32063482
CAS Number: 2154342-25-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR