1-(4-Methylpiperidin-4-yl)ethanone hydrochloride 97%
Supplier: Ambeed
|
Warning
|
A1448583-100MG
A1448583-250MG
76661-330EA
192.03
USD
76661-330
76661-332
1-(4-Methylpiperidin-4-yl)ethanone hydrochloride 97%
1-(4-Methylpiperidin-4-yl)ethanone hydrochloride
Precaución: for Research Use Only
Formula:
C₈H₁₆ClNO MW: 177.6700 g/mol |
CAS Number:
2566476-29-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...