Methyl 4-bromo-2-(bromomethyl)-5-fluorobenzoate 97%

Supplier: Ambeed
Danger

A1448573-100MG A1448573-1G A1448573-250MG
76661-324EA 304 USD
76661-324 76661-326 76661-328
Methyl 4-bromo-2-(bromomethyl)-5-fluorobenzoate 97%
Methyl 4-bromo-2-(bromomethyl)-5-fluorobenzoate

Precaución: for Research Use Only

Formula: C₉H₇Br₂FO₂
MW: 325.9571 g/mol
MDL Number: MFCD32190794
CAS Number: 1614234-12-6
UN: 3261
ADR: 8 Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR