[1,1'-Biphenyl]-4-yl(p-tolyl)methanone 98%

Supplier: Ambeed
Warning

A1448034-100G A1448034-25G
76661-312EA 27.59 USD
76661-312 76661-314
[1,1'-Biphenyl]-4-yl(p-tolyl)methanone 98%
[1,1'-Biphenyl]-4-yl(p-tolyl)methanone

Precaución: for Research Use Only

Formula: C₂₀H₁₆O
MW: 272.3405 g/mol
MDL Number: MFCD00623207
CAS Number: 39148-55-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR