N,N-Diethyl-1-(methylsulfonyl)methanamide 95%

Supplier: Ambeed
Warning

Synonyms: S-Methyl N,N-diethylthiocarbamate sulphone

A1446467-100MG A1446467-1G A1446467-250MG
76661-306EA 98 USD
76661-306 77874-896 77874-897
N,N-Diethyl-1-(methylsulfonyl)methanamide 95%
N,N-Diethyl-1-(methylsulfonyl)methanamide

Precaución: for Research Use Only

Formula: C₆H₁₃NO₃S
CAS Number: 155514-79-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR