2-(4-(Trifluoromethyl)phenyl)-1,3,2-dioxaborinane 98%

Supplier: Ambeed
Warning

A1445714-1G A1445714-250MG
76661-296EA 63.66 USD
76661-296 76661-298
2-(4-(Trifluoromethyl)phenyl)-1,3,2-dioxaborinane 98%
2-(4-(Trifluoromethyl)phenyl)-1,3,2-dioxaborinane

Precaución: for Research Use Only

Formula: C₁₀H₁₀BF₃O₂
MW: 229.9914 g/mol
CAS Number: 416839-38-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR