Potassium-4-methyl-β-styryltrifluoroborate 98%

Supplier: Ambeed
Warning

Synonyms: Potassium (E)-trifluoro(4-methylstyryl)borate

A1445321-1G A1445321-250MG A1445321-5G
76661-286EA 105 USD
76661-286 76661-288 76661-290
Potassium-4-methyl-β-styryltrifluoroborate 98%
Potassium-4-methyl-beta-styryltrifluoroborate

Precaución: for Research Use Only

Formula: C₉H₉BF₃K
MDL Number: MFCD02093982
CAS Number: 219718-86-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR