(R)-3H-Spiro[benzofuran-2,4'-piperidin]-3-amine dihydrochloride 98%

Supplier: Ambeed
Warning

A1445122-100MG A1445122-1G A1445122-250MG
76661-278EA 57 USD
76661-278 76661-280 76661-282
(R)-3H-Spiro[benzofuran-2,4'-piperidin]-3-amine dihydrochloride 98%
(R)-3H-Spiro[benzofuran-2,4'-piperidin]-3-amine dihydrochloride

Precaución: for Research Use Only

Formula: C₁₂H₁₈Cl₂N₂O
CAS Number: 2377356-06-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR