4-Chloro-7-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1-(2,2,2-trifluoroethyl)-1H-indazol-3-amine 98%

Supplier: Ambeed
Warning

A1445117-100MG A1445117-1G A1445117-250MG A1445117-5G
76661-272EA 107.16 USD
76661-272 76661-274 76661-276 77566-918
4-Chloro-7-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1-(2,2,2-trifluoroethyl)-1H-indazol-3-amine 98%
4-Chloro-7-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1-(2,2,2-trifluoroethyl)-1H-indazol-3-amine

Precaución: For research use only.

Formula: C₁₅H₁₈BClF₃N₃O₂
MW: 375.58 g/mol
MDL Number: MFCD34471206
CAS Number: 2189684-53-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR