1,4-Dimethylpyridinium iodide 98%

Supplier: Ambeed
Warning

A1444306-1G A1444306-5G A1444306-25G
76661-258EA 13 USD
76661-258 76661-260 77874-876
1,4-Dimethylpyridinium iodide 98%
1,4-Dimethylpyridinium iodide

Precaución: for Research Use Only

Formula: C₇HIN
MDL Number: MFCD00031899
CAS Number: 2301-80-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR