(5aS,10bR)-2-(3,5-Bis(trifluoromethyl)phenyl)-4,5a,6,10b-tetrahydroindeno[2,1-b][1,2,4]triazolo[4,3-d][1,4]oxazin-2-ium tetrafluoroborate 98%
Supplier: Ambeed
|
Danger
|
A1444277-100MG
A1444277-250MG
76661-254EA
190.97
USD
76661-254
76661-256
(5aS,10bR)-2-(3,5-Bis(trifluoromethyl)phenyl)-4,5a,6,10b-tetrahydroindeno[2,1-b][1,2,4]triazolo[4,3-d][1,4]oxazin-2-ium tetrafluoroborate 98%
(5aS,10bR)-2-(3,5-Bis(trifluoromethyl)phenyl)-4,5a,6,10b-tetrahydroindeno[2,1-b][1,2,4]triazolo[4,3-d][1,4]oxazin-2-ium tetrafluoroborate
Precaución: for Research Use Only
Formula:
C₂₀H₁₄BF₁₀N₃O MW: 513.14 g/mol |
CAS Number:
1214711-40-6 UN: 1759 ADR: 8,Ⅲ |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...