Sodium 2'-(dicyclohexylphosphino)-2,6-diisopropyl-[1,1'-biphenyl]-4-sulfonate 97%

Supplier: Ambeed
Warning

A1443923-100MG A1443923-250MG
76661-242EA 594.11 USD
76661-242 76661-244
Sodium 2'-(dicyclohexylphosphino)-2,6-diisopropyl-[1,1'-biphenyl]-4-sulfonate 97%
Sodium 2'-(dicyclohexylphosphino)-2,6-diisopropyl-[1,1'-biphenyl]-4-sulfonate

Precaución: for Research Use Only

Formula: C₃₀H₄₂NaO₃PS
MW: 536.68 g/mol
MDL Number: MFCD32200884
CAS Number: 870245-84-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR