Methyl-2-chloro-L-mandelate 97%

Supplier: Ambeed
Warning

Synonyms: 2-Chloro-L-mandelic Acid Methyl Ester, (S)-Methyl 2-(2-chlorophenyl)-2-hydroxyacetate, (S)-2'-Chloro-alpha-hydroxyphenylacetic Acid Methyl Ester, Methyl (S)-2'-Chloro-alpha-hydroxyphenylacetate

A1443919-250MG A1443919-5G
76661-238EA 11 USD
76661-238 76661-240
Methyl-2-chloro-L-mandelate 97%
Methyl-2-chloro-L-mandelate

Precaución: for Research Use Only

Formula: C₉H₉ClO₃
MDL Number: MFCD02259731
CAS Number: 32345-60-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR