((2-Fluoro-6-(methoxymethoxy)-8-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)naphthalen-1-yl)ethynyl)triisopropylsilane 97%

Supplier: Ambeed
Warning

A1443865-1G A1443865-250MG A1443865-5G A1443865-25G A1443865-10G
76661-232EA 66.84 USD
76661-232 76661-234 76661-236 77549-750 77566-914
((2-Fluoro-6-(methoxymethoxy)-8-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)naphthalen-1-yl)ethynyl)triisopropylsilane 97%
((2-Fluoro-6-(methoxymethoxy)-8-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)naphthalen-1-yl)ethynyl)triisopropylsilane

Precaución: For research use only.

Formula: C₂₉H₄₂BFO₄Si
MW: 512.5363 g/mol
CAS Number: 2621932-37-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR