tert-Butyl ((2,6-dichloropyridin-3-yl)methyl)carbamate 97%

Supplier: Ambeed
Warning

A1443804-1G A1443804-250MG A1443804-5G
76661-226EA 263.11 USD
76661-226 76661-228 76661-230
tert-Butyl ((2,6-dichloropyridin-3-yl)methyl)carbamate 97%
tert-Butyl ((2,6-dichloropyridin-3-yl)methyl)carbamate

Precaución: for Research Use Only

Formula: C₁₁H₁₄Cl₂N₂O₂
MW: 277.1471 g/mol
MDL Number: MFCD34178753
CAS Number: 2703752-40-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR