2-Chloro-3-fluoro-5-(trifluoromethyl)pyridine 96%
Supplier: Ambeed
|
Danger
|
A1443772-100MG
A1443772-1G
76661-222EA
51.99
USD
76661-222
76661-224
2-Chloro-3-fluoro-5-(trifluoromethyl)pyridine 96%
2-Chloro-3-fluoro-5-(trifluoromethyl)pyridine
Precaución: for Research Use Only
Formula:
C₆H₂ClF₄N MW: 199.533 g/mol |
MDL Number:
MFCD13185448 CAS Number: 72600-67-0 UN: 1993 ADR: 3,Ⅱ |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...