Methyl 5-amino-4,6-dichloropicolinate 98%

Supplier: Ambeed
Warning

A1443591-100MG A1443591-1G A1443591-250MG A1443591-50MG
76661-190EA 165.51 USD
76661-190 76661-192 76661-194 76661-196
Methyl 5-amino-4,6-dichloropicolinate 98%
Methyl 5-amino-4,6-dichloropicolinate

Precaución: for Research Use Only

Formula: C₇H₆Cl₂N₂O₂
MW: 221.0407 g/mol
MDL Number: MFCD28722236
CAS Number: 1805930-73-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR