4-(6-Methyl-1,2,4,5-tetrazin-3-yl)benzoic acid 97%

Supplier: Ambeed
Warning

A1443547-100MG A1443547-1G A1443547-250MG A1443547-50MG
76661-176EA 152.77 USD
76661-176 76661-178 76661-180 76661-182
4-(6-Methyl-1,2,4,5-tetrazin-3-yl)benzoic acid 97%
4-(6-Methyl-1,2,4,5-tetrazin-3-yl)benzoic acid

Precaución: for Research Use Only

Formula: C₁₀H₈N₄O₂
MW: 216.1961 g/mol
MDL Number: MFCD30469092
CAS Number: 1345866-66-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR