3,3'-((2-Aminoethyl)azanediyl)bis(propan-1-ol) 97%

Supplier: Ambeed
Danger

A1443510-100MG A1443510-1G A1443510-250MG
76661-170EA 195.21 USD
76661-170 76661-172 76661-174
3,3'-((2-Aminoethyl)azanediyl)bis(propan-1-ol) 97%
3,3'-((2-Aminoethyl)azanediyl)bis(propan-1-ol)

Precaución: for Research Use Only

Formula: C₈H₂₀N₂O₂
MW: 176.2566 g/mol
MDL Number: MFCD20696639
CAS Number: 50331-68-5
UN: 2735
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR