Methyl 4,5-dibromo-3-chlorothiophene-2-carboxylate 97%

Supplier: Ambeed
Warning

A1443422-100MG A1443422-1G A1443422-250MG A1443422-5G
76661-156EA 42.44 USD
76661-156 76661-158 76661-160 76661-162
Methyl 4,5-dibromo-3-chlorothiophene-2-carboxylate 97%
Methyl 4,5-dibromo-3-chlorothiophene-2-carboxylate

Precaución: for Research Use Only

Formula: C₆H₃Br₂ClO₂S
MW: 334.4128 g/mol
MDL Number: MFCD34178751
CAS Number: 1501789-47-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR