2-(3-(Benzyloxy)propyl)propane-1,3-diol 97%

Supplier: Ambeed
Warning

A1443396-100MG A1443396-1G A1443396-250MG
76661-150EA 18.04 USD
76661-150 76661-152 76661-154
2-(3-(Benzyloxy)propyl)propane-1,3-diol 97%
2-(3-(Benzyloxy)propyl)propane-1,3-diol

Precaución: for Research Use Only

Formula: C₁₃H₂₀O₃
MW: 224.2961 g/mol
MDL Number: MFCD31635054
CAS Number: 132970-27-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR