4-Chloro-2-fluoro-6-methylbenzonitrile 97%
Supplier: Ambeed
|
Danger
|
A1443314-100MG
A1443314-250MG
A1443314-1G
76661-134EA
80
USD
76661-134
76661-136
77874-849
4-Chloro-2-fluoro-6-methylbenzonitrile 97%
4-Chloro-2-fluoro-6-methylbenzonitrile
Precaución: for Research Use Only
Formula:
C₈H₅ClFN |
MDL Number:
MFCD28751903 CAS Number: 1805454-71-0 UN: 3439 ADR: 6.1,Ⅲ |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...