3-Chloro-2-fluoro-6-methylbenzonitrile 97%

Supplier: Ambeed
Danger

A1443312-100MG A1443312-1G A1443312-250MG
76661-128EA 76.39 USD
76661-128 76661-130 76661-132
3-Chloro-2-fluoro-6-methylbenzonitrile 97%
3-Chloro-2-fluoro-6-methylbenzonitrile

Precaución: for Research Use Only

Formula: C₈H₅ClFN
MW: 169.5834 g/mol
MDL Number: MFCD28751897
CAS Number: 1807116-67-1
UN: 3439
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR