N-(4-(2,4-Dihydroxyphenyl)thiazol-2-yl)isobutyramide 97%
Supplier: Ambeed
|
Warning
|
A1443310-1G
A1443310-25G
A1443310-5G
A1443310-250MG
A1443310-100MG
76661-122EA
26
USD
76661-122
76661-124
76661-126
77622-114
77874-848
N-(4-(2,4-Dihydroxyphenyl)thiazol-2-yl)isobutyramide 97%
N-(4-(2,4-Dihydroxyphenyl)thiazol-2-yl)isobutyramide
Precaución: for Research Use Only
Formula:
C₁₃H₁₄N₂O₃S |
CAS Number:
1428450-95-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...