3-Fluoro-2-iodo-5-methylaniline 97%

Supplier: Ambeed
Warning

A1443302-100MG A1443302-10G A1443302-1G A1443302-250MG A1443302-5G
76661-112EA 34 USD
76661-112 76661-114 76661-116 76661-118 76661-120
3-Fluoro-2-iodo-5-methylaniline 97%
3-Fluoro-2-iodo-5-methylaniline

Precaución: for Research Use Only

Formula: C₇H₇FIN
MDL Number: MFCD31707052
CAS Number: 1510866-71-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR