3,5-Difluoro-2-iodoaniline 97%

Supplier: Ambeed
Warning

A1443300-100MG A1443300-10G A1443300-1G A1443300-250MG A1443300-5G
76661-102EA 40.32 USD
76661-102 76661-104 76661-106 76661-108 76661-110
3,5-Difluoro-2-iodoaniline 97%
3,5-Difluoro-2-iodoaniline

Precaución: for Research Use Only

Formula: C₆H₄F₂IN
MW: 255.0039 g/mol
MDL Number: MFCD28789178
CAS Number: 1804885-53-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR