N-(3,6-Dichloropyridazin-4-yl)acetamide 97%

Supplier: Ambeed
Warning

A1443290-100MG A1443290-1G A1443290-250MG
76661-092EA 17 USD
76661-092 76661-094 76661-096
N-(3,6-Dichloropyridazin-4-yl)acetamide 97%
N-(3,6-Dichloropyridazin-4-yl)acetamide

Precaución: for Research Use Only

Formula: C₆H₅Cl₂N₃O
MDL Number: MFCD09865627
CAS Number: 121163-50-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR