Diethylene Glycol Mono(2-propyn-1-yl) ether 95%

Supplier: Ambeed
Warning

Synonyms: 2-[2-(2-Propynyloxy)ethoxy]ethanol, 2-[2-(2-Propyn-1-yloxy)ethoxy]ethanol, Propargyl-PEG2-OH

A1443008-1G A1443008-250MG A1443008-25G A1443008-5G
76661-076EA 20 USD
76661-076 76661-078 76661-080 76661-082
Diethylene Glycol Mono(2-propyn-1-yl) ether 95%
Diethylene Glycol Mono(2-propyn-1-yl) ether

Precaución: for Research Use Only

Formula: C₇H₁₂O₃
MW: 144.17 g/mol
MDL Number: MFCD00053353
CAS Number: 7218-43-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR