1,6-Bis(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrene 98%

Supplier: Ambeed
Warning

A1442340-100MG A1442340-1G A1442340-250MG A1442340-5G
76661-068EA 39 USD
76661-068 76661-070 76661-072 76661-074
1,6-Bis(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrene 98%
1,6-Bis(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrene

Precaución: for Research Use Only

Formula: C₂₈H₃₂B₂O₄
MDL Number: MFCD32880071
CAS Number: 950779-13-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR