2,7-Dimethoxy-9H-thioxanthen-9-one 98%

Supplier: Ambeed
Warning

A1442334-250MG A1442334-5G
76661-064EA 45.62 USD
76661-064 76661-066
2,7-Dimethoxy-9H-thioxanthen-9-one 98%
2,7-Dimethoxy-9H-thioxanthen-9-one

Precaución: for Research Use Only

Formula: C₁₅H₁₂O₃S
MW: 272.3190 g/mol
MDL Number: MFCD33404409
CAS Number: 435344-88-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR